Beskyttelsesbriller, Flex Seal™
Leverandør: HONEYWELL SAFETY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1011333 1011381
111-0132EA 300 DKK
111-0132 111-3606
Beskyttelsesbriller, Flex Seal™
Beskyttelsesbriller
Goggles with a flexible and sturdy frame feature a wraparound lens for a panoramic view.
  • Soft and flexible silicone frame for excellent comfort and ensures a snug, slip-free fit
  • Remains soft and supple even in extremely hot or cold environments
  • Pivoting, removable and adjustable headband
  • Indirect ventilation prevents fogging
  • Neoprene strap (111-0132) or elastic strap (111-3606)
Designed to be compatible with prescription spectacles.
Vægt: 110 g.
Certificeringer: EN 166, EN 169, EN 171, 1FT, C 3, 4, 9.