Industrial gloves, Hylite® 47-400 / 47-402 / 47-409
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
47-400/8 47-400/9 47-409/8 47-409/9 47-400/7 47-402/9 47-402/10 47-402/7 47-402/8 47-409/10 47-409/7 47-400/10
112-0292EA 459 DKK
112-0292 112-0294 112-3706 112-0295 112-0291 ANSE47-402/9 ANSE47-402/10 ANSE47-402/7 ANSE47-402/8 112-3708 112-3705 112-4760
Industrial gloves, Hylite® 47-400 / 47-402 / 47-409
Handsker
The 100% interlock knit liner features a nitrile coating on the entire glove (47 - 402/47 - 409) or on the palm only (47 - 400). The glove is silicone-free and machine washable.
  • Superior resistance to cut, puncture and abrasion with excellent flexibility and dexterity
  • Repels grease and oil
  • 100% cotton interlock knit liner is comfortable and non irritating
  • Ergonomical design and generous sizing improves dexterity and reduces hand fatigue
  • Textured finish for an outstanding wet and dry grip even on the smoothest surface
The standard models feature a knitted wrist, the Nitragold™ model has a nitrile coated wrist.
Certificeringer: EN 388:2016 (3111A); Cat. II