Heat resistant gloves, ActivArmr® 43-113
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
43-113/10 43-113/11
112-0290EA 591 DKK
112-0290 112-0449
Heat resistant gloves, ActivArmr® 43-113
Handsker
These ambidextrous Kevlar® gloves are suitable for use in the most exacting hot applications. They also provide fantastic resistance to cuts, tears and abrasion.
  • Pure cotton liners, comfortable and cool to wear
  • No seams where sparks or metal splashes might lodge
  • May be repeatedly laundered and re-used with no loss of performance
  • Designed for intermittent dry heat contact up to 350 °C
Certificeringer: EN 388:2016 (154XE), EN 407 (43421X); Cat. III