Industrial gloves, Perfect Poly® white
Leverandør: HONEYWELL SAFETY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2232255/10 2232255/6 2232255/7 2232255/9 2232255/8
DALL2232255/10EA 0 DKK
DALL2232255/10 DALL2232255/6 DALL2232255/7 DALL2232255/9 DALL2232255/8
Industrial gloves, Perfect Poly® white
Handsker
Knitted gloves made of lightweight, white polyamide with PU coating. Suitable for the handling of fine products in a dry or slightly greasy or soiled environment.
  • Polyurethane coating on palm and fingertips
  • Exceptional dexterity
  • High abrasion resistance
  • Non pilling: Suitable for precision work
  • Certified as 'silicone-free' to ensure their use in painting environment
Typical applications include: Automotive industry assembly, logistics (forklift operators), and sorting of parts.
Certificeringer: EN 388:2016 (3131X); Cat. II