Chemical resistant gloves, AlphaTec® 79-700
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
79-700/10 79-700/11 79-700/7 79-700/8 79-700/9
ANSE79-700/10EA 282 DKK
ANSE79-700/10 ANSE79-700/11 ANSE79-700/7 ANSE79-700/8 ANSE79-700/9
Chemical resistant gloves, AlphaTec® 79-700
Handsker
This nitrile glove is designed specifically to offer outstanding comfort, flexibility and good chemical resistance.
  • Extremely thin and soft, offers the dexterity of a single use glove
  • Specific lozenge pattern increases oil grip
  • Non chlorinated outer surface helps to enhance grip
  • Ansell Moisture Management Technology™: lower tendency to shed (even up to 50% less shedding) compared to traditional cotton flock gloves
The Aquadri™ technology (Ansell Moisture Management Technology™) gives this reusable glove excellent moisture absorption capabilities and keeps users hands drier for a longer period of time. The nitrile formulation is designed to provide good mechanical and chemical performance levels especially in environments where hydro-carbon based chemicals, oils and greases are present.
Thickness: 0,225 mm
Colour: Blue
EN 374, EN 388 (1001), EN 421, EN 1149; AQL 0.65
Certificeringer: EN ISO 374-1:2016 Type B (JKT), EN ISO 374-5 VIRUS, EN 388:2016 (1001A), EN 421; Cat. III
Food contact approved