Kemisk resistente handsker, North Butyl™
Leverandør: Honeywell Safety
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
B131/10 B131/7 B131/8 B131/9 B131R/8 B131R/10 B131R/11 B131R/7 B131R/9
NORHB131/10EA 1190 DKK
NORHB131/10 NORHB131/7 112-2542 112-2543 NORHB131R/8 NORHB131R/10 NORHB131R/11 NORHB131R/7 NORHB131R/9
Kemisk resistente handsker, North Butyl™
Handsker
These unsupported butyl gloves provide the highest permeation resistance to gas and water vapor. They are highly resistant to ketones (MEK, MIBL, acetone) and esters (tricresyl phosphate, amyl acetate, ethyl acetate).
  • Flexible and sensitive, even at lower temperatures
  • Remains supple from −30 to +50 °C
  • Curved fingers and hand-specific design provide a comfortable fit
  • Rolled cuffs for easy donning and removal
Available in smooth finish or with Grip-Saf™ palm for enhanced grip.
Thickness: 0,33 mm
Colour: Black
EN 374 (BCI)
Certificeringer: EN 388 (2000), EN 374 (BCI); Cat. III