Kemikaliebestandige handsker, UltraNeo 401
Leverandør: MAPA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
34401140 34401147 34401148 34401149
112-0679EA 57.5 DKK
112-0679 112-0681 112-0682 112-0683
Kemikaliebestandige handsker, UltraNeo 401
Handsker
Neopren/naturlatex handsker med flok bomuldsfor.
  • Tactile sensitivity for light chemical protection
  • Added comfort thanks to the cotton flock lining
  • Great dexterity, due to the glove’s reduced thickness
  • Embossed textures for good grip on slippery objects
Ideal for use in the automotive/mechanical industry, for battery manufacturing, the preparation of industrial glues and industrial cleaning.
Thickness: 0,55 or 0,75 mm
Colour: Black
EN 388, EN 374 (AKL); Cat. III
Certificeringer: EN 388:2016 (2110X), EN ISO 374-1 Type A (ALMNST), EN ISO 374-5; Cat. III