Examination gloves, neoprene, Micro-Touch® Affinity™
Leverandør: ANSELL MEDICAL PRODUCTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
553355 553354 553353 553352
112-1828EA 295 DKK
112-1828 112-1827 112-1826 112-1825
Examination gloves, neoprene, Micro-Touch® Affinity™
Handsker
These gloves are designed to be worn on either hand and are free of latex proteins, providing outstanding protection from type I allergies for medical staff and patients. They are designed with a beaded cuff and textured fingertips, providing a secure grip even in damp conditions.
  • Non sterile, powder-free, textured fingertips
  • Polyacrylate inner coating for easy donning
  • Outstanding flexibility and elongation offer a level of comfort comparable to latex
  • Highly resistant to a wide range of chemicals
  • Better puncture resistance than latex gloves