Nitrile gloves, Solo Ultra 997
Leverandør: MAPA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
34997756 34997757 34997758 34997759
112-0771EA 264 DKK
112-0771 112-0772 112-0773 112-0774
Nitrile gloves, Solo Ultra 997
Handsker
These ambidextrous liquidproof gloves with beaded cuffs are made of nitrile.
  • Non sterile, chlorinated, textured fingertips
  • Excellent dexterity thanks to flexibility and reduced material thickness
  • Easy to put on and take off
  • Protection of the hand as well as handled products
  • Recommended for people sensitised to natural rubber proteins
Ideal for food handling, laboratory work, small parts assembly.
Certificeringer: EN ISO 374-1 Type B (JKT), EN ISO 374-5 VIRUS, Cat. III; AQL 1,5
Food contact approved.