Glass cutter
Leverandør: USBECK Laborgeräte
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5210 5211
201-9010EA 548 DKK
201-9010 201-9011
Glass cutter
Knive
Plastic handle with hardened metal blade.
  • Clean and precise cutting
  • Replaceable hardened metal blades
  • Easy to change blade
Length 185 mm, blade dimensions 40×20 mm.