Immersion Filters
Leverandør: ROBU GLASFILTER
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
24202 24201 24203 24204 24350 24351 24352 24353 24354 24200
511-2312EA 380 DKK
511-2312 511-2311 511-2313 511-2314 ROBU24350 ROBU24351 ROBU24352 ROBU24353 ROBU24354 511-2310
Immersion Filters
Filtre Filterrør
ROBU® immersion filters with VitraPOR® glass filter are designed for efficient reverse filtration of liquids. Ideal for applications requiring fine particle separation directly within the vessel.
  • For reverse filtration
  • Integrated VitraPOR® filter
  • Reusable, chemical resistant
  • Suitable for laboratory and process use
  • Various porosities available
  • Easy immersion into vessels
  • Material: Borosilicate glass 3.3
ROBU immersion filters equipped with a VitraPOR glass filter enable highly efficient reverse filtration directly within containers or vessels. Designed for laboratory and industrial applications, these filters are ideal when a vacuum or pressure-driven reverse flow is required.
The chemically resistant borosilicate glass ensures durability, while the precision of the VitraPOR element guarantees consistent separation of fine particles. These reusable filters come in a variety of porosities and lengths, providing flexibility for a wide range of filtration tasks.
Certificeringer:
In compliance with ISO 4796-80 and ASTM E128-99 standards.