Dropping funnels, pear shaped, with pressure balance, Quickfit®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
DE100/22 DE250/32 DE250/33
201-8361EA 2210 DKK
201-8361 201-8363 201-8364
Dropping funnels, pear shaped, with pressure balance, Quickfit®
Tragte Dråbetragte
PYREX®, borosilicate glass.
  • Rotaflo® stopcock features chemically resistant PTFE keys
  • High chemical and temperature resistance
Pear shaped, with standard ground joint socket and Rotaflo® stopcock. Used when liquids have to be introduced into vessels under vacuum.
Cone sizes are under Joint size (NS) column.
Certificeringer: DIN ISO 4800