Conical separating funnels
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2190/02M 2190/04M 2190/06M 2190/08M 2190/10M 2190/01M
511-0760EA 963 DKK
511-0760 511-0761 511-0762 511-0763 511-0764 532-0103
Conical separating funnels
Tragte Skilletragte
PYREX®, borosilicate glass, with glass stopper.
With ground glass stopper and interchangeable PTFE key. Without graduation.