Koniske skilletragte
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
04000028 04000037 04000049 04000058 04000070 04000073 04002073 04004170 04002028 04002037 04002049 04002058 04002070 04004128 04004137 04004149 04004158 04004173 04006128 04006137 04006149 04006158 04006170 04006173
527-1131EA 447 DKK
527-1131 527-1132 527-1133 527-1224 527-1134 527-1135 527-1226 527-1140 527-1221 527-1222 527-1223 527-1214 527-1225 527-1136 527-1137 527-1138 527-1139 527-1141 527-1201 527-1202 527-1203 527-1204 527-1205 527-1206
Koniske skilletragte
Tragte Skilletragte
DURAN®, borosilikatglas, med NS-PE prop.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. Both with or without graduations.