Cylinderformede skilletragte
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
04022049 04022058 04022028 04022037 04022070 04026128 04026137 04026149 04026158 04026170
527-1123EA 623 DKK
527-1123 527-1124 527-1121 527-1122 527-1125 527-1101 527-1102 527-1103 527-1104 527-1105
Cylinderformede skilletragte
Tragte Skilletragte
DURAN®, borosilikatglas, med NS-PE prop.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. With graduation.