Funnel
Leverandør: Sartorius
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6981002 6981065
516-7033EA 3410 DKK
516-7033 516-7034
Funnel
Tragte Membranfiltertragte
Rustfrit stål, autoklaverbar.
To use with filter holders for colony counting.
Bestillingsoplysninger: Supplied with funnel and clamp (lid and seal must be ordered seperately).