Skilletragte, cylinderformede, uden trykudligning
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
04172814 04174914 04175814 04177014 04172811 04173711 04173714 04172211
201-1125EA 730 DKK
201-1125 201-1127 201-1128 201-1129 201-1124 271-5123 201-1126 LENZ04172211
Skilletragte, cylinderformede, uden trykudligning
Tragte Dråbetragte
DURAN®, borosilicate glass, clear, with PTFE key.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Cylindrical shape. With standard ground joint cone and socket. Clearly visible white divisions for approximate volume reading.
DIN ISO 4800
Certificeringer: DIN ISO 4800