Funnel for conical ground joints
Leverandør: VITLAB
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
70594 70694 70494
221-1409EA 573 DKK
221-1409 221-1410 221-1418
Funnel for conical ground joints
Tragte Filtreringstragte Tragte til konisk slebne samlinger
PP, translucent.
  • Ideal for transferring liquid or powdered reagents to multiple-neck flasks
  • Very good chemical resistance
  • Autoclavable
The funnel body is flattened on the side and the end of the shaft is shaped to fit conical ground joints.