Analytical funnel
Leverandør: Kavalier
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
632421052055 632421052075 632421052100
221-1715EA 396 DKK
221-1715 221-1716 221-1717
Analytical funnel
Tragte Filtreringstragte Analysetragte
Borosilikatglas 3,3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Til hurtig filtrering.