Funnel stands for two or four funnels, LaboPlast®
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9622-0400 9622-0200
519-2722EA 2340 DKK
519-2722 519-2721
Funnel stands for two or four funnels, LaboPlast®
Retortstativer
PP, chrome-plated steel. Sturdy and stable funnel stand, resistant to chemicals, shatter-proof and with continuously variable height adjustment.
  • Funnel stand is suitable for all funnels from 40 to 180 mm in diameter
  • Funnel holders can be used with reducing inserts for small funnels
  • Funnel stand also makes an excellent filtering stand
  • Adjustable height up to 450 mm
  • Additional funnel holders are available as accessories
Thanks to an innovative magnet concept no clamping screws are necessary. The height of the funnel holder can be adjusted easily andMore Product Information