Forceps with guide pin
Leverandør: BOCHEM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1810 1811 1812
232-0016EA 332 DKK
232-0016 232-2137 232-0017
Forceps with guide pin
Pincet
Stainless steel, PTFE coating.
  • Anti-magnetic
  • Very good chemical and thermal resistance
  • Smooth surface prevents contamination and facilitates cleaning