Tweezers
Leverandør: VITLAB
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
68299 68399 68099 68199
232-1951EA 40.9 DKK
232-1951 232-1953 VITL68099 VITL68199
Tweezers
Pincet
POM, yellow. Universal tweezers for a variety of applications.
  • Blunt, elastic, very good resilience
  • With grooves on the outside for optimum handling and grip
  • Good chemical resistance