Forceps, precision
Leverandør: OUTILS RUBIS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
44 3 3C 5 7 5B 2 1 4 35A 35B AA F 6 0 SS-SA
232-1132EA 221 DKK
232-1132 232-1219 232-1220 232-1221 232-1230 OUTI5B 232-0000 232-0143 232-0216 232-0209 OUTI35B 232-0217 OUTIF OUTI6 OUTI0 OUTISS-SA
Forceps, precision
Pincet
Stainless steel. Forceps with very precisely machined tips. Ideal for applications that require a high degree of precision, including microscopy, anatomy, biology, microbiology, gene technology and electron microscopy.
  • Acid-resistant
  • Resistant against hydrogen fluoride and nitric acid
  • 100% anti-magnetic