Målekolber, Klasse A
Leverandør: HIRSCHMANN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2820392 2820393 2820394 2820397 2820398 2820366 2820381 2820361 2820380 2820353 2820360 2820390 2820352 2820365 2820370 2820371 2820375 2820378 2820385 2820386
612-3386EA 520 DKK
612-3386 612-3387 612-3502 612-3503 HIRS2820398 HIRS2820366 HIRS2820381 HIRS2820361 HIRS2820380 HIRS2820353 HIRS2820360 HIRS2820390 HIRS2820352 HIRS2820365 HIRS2820370 HIRS2820371 HIRS2820375 HIRS2820378 HIRS2820385 HIRS2820386
Målekolber, Klasse A
Kolber Volumetriske kolber
DURAN®, borosilicate glass, without stopper, conformity certified (DE-M marking).
  • Blue enamelled indelible markings resistant to acids and alkalis
  • Certificate of conformity with dated batch identification available via website: (www.hirschmannlab.de)
*Wide neck
Certificeringer: DIN EN ISO 1042