Målekolber, Klasse A
Leverandør: HIRSCHMANN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2640152 2640153 2640160 2640161 2640165 2640170 2640175 2640178 2640180 2640181 2640185 2640186 2640190 2640193 2640194 2640171
612-1539EA 563 DKK
612-1539 612-1540 612-1541 612-1542 612-1543 612-1544 612-1545 612-1546 612-1547 612-1548 612-1549 612-1551 612-1552 612-1553 612-1554 612-4715
Målekolber, Klasse A
Kolber Volumetriske kolber
DURAN®, borosilicate glass, amber, with PE stopper, conformity certified (DE-M marking).
  • White enamel indelible markings resistant to acids and alkalis
  • Certificate of conformity with dated batch identification available via website: (www.hirschmannlab.de)
*Wide neck.
Certificeringer: DIN EN ISO 1042