Volumetric flasks, class A or B, blue markings
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
36162 36178 36182
612-3796EA 282 DKK
612-3796 612-4810 612-4812
Volumetric flasks, class A or B, blue markings
Kolber Volumetriske kolber
PMP, transparent, with PP stopper.
  • Calibrated to contain (TC, In)
  • Ring mark and inscriptions in blue
  • Exposure to temperature up to 121 °C (autoclaving) will not cause permanently exceeded error limits! Cleaning up to 60 °C is recommended to preserve marks and inscriptions
Error limits according to class A respectively B, DIN EN ISO 1042.
*Bred hals