Volumetric flasks, class B
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
36020 36028 36038 36048 36054 36062 36008
612-4076EA 111 DKK
612-4076 612-4077 612-4078 612-4079 612-4080 612-4081 612-3708
Volumetric flasks, class B
Kolber Volumetriske kolber
High clarity PP, with PP stopper.
  • Calibrated to contain (TC, In)
  • Ring mark and inscriptions in blue
  • Exposure to temperatures up to +60 °C (autoclaving) will not cause permanently exceeded error limits, Cleaning up to max. +60 °C is recommended to preserve marks and inscriptions
Error limits according to class B, DIN EN ISO 1042.
*Bred hals.