Målekolber, Klasse B, Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4001-0050 4001-0100 4001-0250 4001-0500 4001-1000
612-3021EA 393 DKK
612-3021 612-3022 612-3024 612-3025 612-3026
Målekolber, Klasse B, Nalgene®
Kolber Volumetriske kolber
PMP (TPX®), transparent, with PP screw cap.
  • Very robust and allowing precise measurement
  • Calibrated individually with permanent colour graduation line
  • Autoclavable when the stopper has been removed from the threaded neck and a small quantity of water has been added to the flask. Cleaning with mild alkaline detergents recommended up to +60 °C (accuracy may be affected over +60 °C)
ASTM E288 and ISO Standard 384 Class B
Certificeringer: ASTM E288 og ISO Standard 384