Målekolber, Klasse B, Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4000-0050 4000-0100 4000-0200 4000-0250 4000-0500 4000-1000
612-3011EA 406 DKK
612-3011 612-3012 612-3013 612-3014 612-3015 612-3016
Målekolber, Klasse B, Nalgene®
Kolber Volumetriske kolber
PPCO, translucent, with PP screw cap.
  • Very robust and allowing precise measurement
  • Calibrated individually with permanent colour graduation line
  • Autoclavable when the stopper has been removed from the threaded neck and a small quantity of water has been added to the flask. Cleaning with mild alkaline detergents recommended up to +60 °C (Accuracy may be affected over +60 °C)
ASTM E288 and ISO Standard 384 Class B
Certificeringer: ASTM E288 og ISO Standard 384