Målekolber, Klasse A
Leverandør: HIRSCHMANN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2640252 2640253 2640260 2640265 2640270 2640275 2640280_U 2640285 2640290_U 2640293 2640294
612-3465EA 399 DKK
612-3465 612-3466 612-3467 612-3469 612-3470 612-3471 612-3473 612-3475 612-3477 612-3478 612-3479
Målekolber, Klasse A
Kolber Volumetriske kolber
DURAN®, borosilicate glass, amber, with glass stopper, conformity certified (DE-M marking).
  • White enamelled indelible markings resistant to acids and alkalis
  • Certificate of conformity with dated batch identification available via website: (www.hirschmannlab.de)
*Wide neck.
Certificeringer: DIN EN ISO 1042.