Målekolber, Klasse A
Leverandør: HIRSCHMANN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2800152 2800160 2800165 2800170 2800175 2800180 2800185 2800186 2800190 2800193 2800194 2800197 2800198
612-3277EA 142 DKK
612-3277 612-3278 612-3279 612-3280 612-3281 612-3282 612-3283 612-3284 612-3285 612-3286 612-3287 612-3288 612-3289
Målekolber, Klasse A
Kolber Volumetriske kolber
DURAN®, borosilicate glass, with beaded rim, conformity certified (DE-M marking).
  • Blue enamelled indelible markings resistant to acids and alkalis
  • Certificate of conformity with dated batch identification available via website: (www.hirschmannlab.de)
* In addition to DIN
Certificeringer: DIN EN ISO 1042