Volumetric flasks, class A, white markings, BLAUBRAND®
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
37411 37401 37402 37403 37405 37406 37407 37408 37409 37410 37404 2.37449 37412 37413
612-5094EA 657 DKK
612-5094 612-5084 612-5085 612-5086 612-5088 612-5089 612-5090 612-5091 612-5092 612-5093 612-5087 612-5736 612-6789 612-6790
Volumetric flasks, class A, white markings, BLAUBRAND®
Kolber Volumetriske kolber
Borosilicate glass 3.3, amber, with PP stopper.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in high contrast white enamel
  • DE-M marking
*Bred hals.
Certificeringer: DIN EN ISO 1042
Bestillingsoplysninger: Supplied with a batch certificate. Volumetric flasks also available on request with an individual certificate, a USP individual certificate or a DAkkS calibration certificate.