Flasks, round bottom, wide neck, with flared rim
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1120/18D 1120/08D 1120/14D 1120/22D 1120/04M
214-0426EA 6480 DKK
214-0426 214-0025 BIBB1120/14D 214-0027 BIBB1120/04M
Flasks, round bottom, wide neck, with flared rim
Kolber Kolber med rund bund
PYREX®, borosilicate glass, wide neck.
  • Meget god kemisk resistens
  • Høj temperaturresistens
Wide neck to facilitate easy addition of solid samples etc.. With white enamel marking spot.
Certificeringer: ISO 1773