Rotary evaporating flasks
Leverandør: BUCHI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
000431 000432 000433 000434 000435 000436 000437 020729 025323 025520
531-1811EA 768 DKK
531-1811 531-1812 531-1813 531-1814 531-1815 531-1816 531-1817 BUCH020729 BUCH025323 BUCH025520
Rotary evaporating flasks
Kolber Kolber til rotationsfordamper
Made of borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For Rotavapor® R-210/215 rotary evaporators. Pear-shaped with NS 29.2/32 ground joint socket.