Flasks, flat bottom, wide neck, with flared rim
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1080/10D 1080/12D 1080/08D
BIBB1080/10DEA 1480 DKK
BIBB1080/10D BIBB1080/12D 511-0733
Flasks, flat bottom, wide neck, with flared rim
Kolber Fladbundede kolber
PYREX®, borosilicate glass, wide mouth.
  • Meget god kemisk resistens
  • Høj temperaturresistens
Wide neck to facilitate easy addition of solid samples etc.. Flat base for standing on benches and hotplates. With white enamel marking spot.
Certificeringer: ISO 1773