Flasks, flat bottom, narrow neck
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1070/18D 1070/24D 1070/28D 1070/32D 1070/36D 1070/38D 1070/04D 1070/12D
214-0346EA 147 DKK
214-0346 214-0347 214-0348 214-0012 214-0013 214-0014 214-0008 BIBB1070/12D
Flasks, flat bottom, narrow neck
Kolber Fladbundede kolber
PYREX®, borosilicate glass, clear.
  • High resistance to thermal shock
  • Excellent chemical durability for contamination-free performance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Narrow neck flasks with flat base for standing on benches and hotplates. Marked with capacity and a white labelling area.
Certificeringer: ISO 1773