Nitrogen receivers (schlenk Tubes)
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
03370149 03370213 03370222 03370237 03370113 03370122 03370128 03370137 03370228 03370249
LENZ03370149EA 672 DKK
LENZ03370149 LENZ03370213 LENZ03370222 LENZ03370237 201-1161 LENZ03370122 LENZ03370128 LENZ03370137 LENZ03370228 LENZ03370249
Nitrogen receivers (schlenk Tubes)
Kolber Nitrogenflasker
DURAN®, borosilikatglas, klar.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With 14/23 standard ground sockets/cones and standard ground side stopcock with tube connector.