Flasks, round-bottomed with two necks and standard ground joints
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
03203137 03204149 03204158 03204170 03204173 03204449 03204458 03204470 03204473 03204479 03204137 03203149 03203158 03203170 03203237 03203249 03203258 03203270
LENZ03203137EA 257 DKK
LENZ03203137 271-1426 271-1428 271-1430 201-1411 271-1427 271-1429 271-1431 LENZ03204473 LENZ03204479 271-1424 LENZ03203149 LENZ03203158 LENZ03203170 LENZ03203237 LENZ03203249 LENZ03203258 LENZ03203270
Flasks, round-bottomed with two necks and standard ground joints
Kolber Kolber med rund bund
DURAN®, borosilicate glass, with parallel side neck.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With standard ground centre neck and matt white labelling area.