Flasks acc. to Kjeldahl with standard ground joint
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
03051937 03052949 03052958 03052964 03051928 03052970
214-0176EA 404 DKK
214-0176 214-0177 LENZ03052958 214-0178 LENZ03051928 LENZ03052970
Flasks acc. to Kjeldahl with standard ground joint
Kolber Kjeldahl-kolber
DURAN®, borosilicate glass, clear.
  • Meget god kemisk resistens
  • Høj temperaturresistens
  • Minimal varmeudvidelse giver relativt høj resistens overfor temperaturændringer
With labelling area.
Certificeringer: DIN 12360