Flasks, flat bottom with Quickfit® ground joint
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
FF250/3S FF50/3S FF100/2S FF500/3S FF250/5S FF100/3S FF100/4S FF150/5S FF1L/4S FF250/4S FF50/2S FF500/4S FF150/3S FF250/2S FF1L/3US FF150/4S FF500/5S FF2L/3S FF2L/4S FF150/2S FF1L/5S FF2L/5S
201-8443EA 977 DKK
201-8443 201-8448 201-8430 201-8449 201-8444 201-8432 201-8034 201-8437 201-8037 214-0048 201-8447 201-8036 201-8436 201-8441 201-8439 BIBBFF150/4S BIBBFF500/5S 214-0417 BIBBFF2L/4S BIBBFF150/2S BIBBFF1L/5S BIBBFF2L/5S
Flasks, flat bottom with Quickfit® ground joint
Kolber Fladbundede kolber
PYREX®, borosilicate glass, clear, with short neck.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Flat base allows use with hotplates. Labelling area.