Vacuum filtering flasks, Büchner, with glass hose connection
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
211916802 211917301 211918803 211919105 212012409 212013602 212014401 212015406 212016308 211918606
519-4217EA 1480 DKK
519-4217 519-4216 511-0218 511-0133 215-1011 215-1013 215-1018 215-1010 519-4140 511-0217
Vacuum filtering flasks, Büchner, with glass hose connection
Kolber Filterkolber
DURAN®, borosilikatglas 3.3, klar.
  • Up to 2000 ml in Erlenmeyer shape, from 3000 ml bottle shape
  • Olive hose connection 11 mm Ø
Til vakuumbrug.
ISO 6556 (only Erlenmeyer shape)
Certificeringer: ISO 6556 (Erlenmeyer-form)
DIN 12475 (Flaskeform)