Replacement impeller for Celstir® flasks
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
356899 356895 356910 356909 356907 356904 356902 356896 356893 356903
734-3162EA 3000 DKK
734-3162 734-3160 734-3168 734-3167 734-3166 734-3165 734-3163 734-3161 734-3159 WHEA356903
Replacement impeller for Celstir® flasks
Omrørere Omrøreaksler
This unit includes a top cap with liners, glass rod, magnet holder, magnet and stirring paddle (for sizes 125 ml or larger).