Erlenmeyer flasks, narrow neck, with flared rim
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1130/36D 1130/10D 1130/03 1130/05M 1130/12D 1130/26D 1130/30D 1130/16D 1130/32D 1130/02M 1130/06D 1130/11M 1130/14D 1130/20D 1130/08D
214-0040EA 848 DKK
214-0040 214-0055 214-0029 214-0030 214-0033 214-0037 214-0038 214-0035 214-0039 214-0028 214-0031 214-0032 214-0034 214-0036 BIBB1130/08D
Erlenmeyer flasks, narrow neck, with flared rim
Kolber Erlenmeyer-kolber
PYREX®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
White graduations and labelling area.
Certificeringer: ISO 1773