Kulstoffiltreringskapsler, Carbon Cap™, Whatman™
Leverandør: Whatman products (Cytiva)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6704-1500 6704-7500
515-0105EA 1550 DKK
515-0105 515-0106
Kulstoffiltreringskapsler, Carbon Cap™, Whatman™
Filtre Ventilationsfiltre
Carbon Cap™ capsule filters are filled with activated carbon and include a pleated glass fiber filter. They are suitable for adsorption of organic vapours from air, and for removing colour, organics, and chlorine from water.
  • Activated carbon acts as an adsorption media for colour, organics, and chlorine
  • Pleated glass microfiber filter structure increases surface area, resulting in high efficiency
  • Retains 99,97% of particles greater than 0,3 µm
Large volume chlorine and organic filtration are facilitated by activated carbon, which adsorbs these molecules.
Carbon Cap™ is a filter filled with high-purity, high-efficiency, acid-washed, activated carbon and a High Efficiency Particulate Air (HEPA) filter. It is made specifically for adsorption of organic vapours, chlorine, and decolorising liquids.