Tangential flow systems, Vivaflow® 50 and 200
Leverandør: Sartorius
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VF20P0 VF20P2 VF20P3 VF20P4 VF20P7 VF20H1 VF20H0 VF20H2 VF20P1 VF20H9 VF20P9 VF20H4
512-4069EA 5360 DKK
512-4069 512-4070 512-4071 512-4072 512-4074 515-0140 515-0139 515-0141 511-0876 518-0090 518-0091 518-0104
Tangential flow systems, Vivaflow® 50 and 200
Filtre Tangentialflowskassetter
Vivaflow® 50 and 200 devices are ready to use crossflow cassettes for filtration and concentration of samples from 100 to 3000 ml or 500 to 5000 ml. The membrane surface per device is 50 cm² or 200 cm² respectively. The devices come with all the necessary accessories for running with a laboratory pump and a size 16 pump head. For speeding up the concentration and increasing the volume throughput, up to six Vivaflow® 50 cassettes can be connected together or two Vivaflow® 200 can be connected together. Optimally suited for applications like cell culture supernatant concentration, virus concentration and water concentration this economical product offers a standard of ease of use, reliability and flexibility.
  • Plug and play laboratory crossflow cassette
  • Optimal for aqueous sample concentration e.g. proteins and nanoparticles
  • Vivaflow® 50 are disposable. Vivaflow® 200 are reusable
  • High flow rate and recovery