Membranfiltre, Whatman™
Leverandør: Whatman products (Cytiva)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
10410012 10410014 10410206 10410212 10410214 10410219 10410224 10410229 10410312 10410314 10410319 10410380
514-2422EA 3120 DKK
514-2422 514-0768 514-2451 514-2452 514-0487 514-2454 514-0488 514-0489 514-2462 514-2463 514-2464 514-0681
Membranfiltre, Whatman™
Filtre Membranfiltre
These hydrophilic regenerated cellulose membrane filters are microporous cellulose acetate filters made without wetting agents. RC60 offers low extractables and wide compatibility in both organic and aqueous media.
  • Spontan hydrofil, meget god vådstyrke
  • Kan steriliseres med alle metoder
Suitable for use as sterile filter to ASTM-D 3862-80