TCLP glasfiberfiltre
Leverandør: Cytiva (Formerly Pall Lab)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
66256 66257 66258 60076
513-0024EA 1160 DKK
513-0024 513-0025 513-0026 PALL60076
TCLP glasfiberfiltre
Filtre Glasfiberfiltre
Lavt niveau af ekstraherbare stoffer, bindemiddelfri borosilikatglasfiberfiltre. Velegnet til brug i EPA 1311 TCLP eller til forfiltrering af urene HPLC-prøver.
  • Reduces filtration costs and premature clogging when filtering difficult-to-filter or highly contaminated solutions
  • Extends filter life and reduces final filter changes with high capacity prefilters
  • Eliminates sample contamination, binder-free borosilicate glass fiber has no added extractables
Filters a wide range of particulate loads and viscous solutions with a selection of filterMore Product Information
Bestil nu
Specifikationer
Environmentally Preferable
Thickness 432 µm 432 µm
Partikelretention 0.7 µm
Porestørrelse 0,7 µm 0,7 µm