Vakuumfiltreringssystemer med flasketop, VacuCap®, Supor®-membran
Leverandør: Cytiva (Formerly Pall Lab)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4621 4622 4624 4632 4634 TA4622 TA4632 4628 4631 4638
515-0012EA 2080 DKK
515-0012 515-0013 515-0014 515-0017 515-0018 516-9812 516-9813 515-0015 515-0016 515-0019
Vakuumfiltreringssystemer med flasketop, VacuCap®, Supor®-membran
Filtre Topfiltre til flasker
Supor® hydrophilic polyethersulphone (PES) membrane; PP housing. VacuCap® filtration device eliminates the need to constantly refill funnel in traditional bottle top filters as it draws directly from the mixing reservoir. The patented small design accepts a variety of collection vessels and reduces laboratory storage space and waste. The Supor® membrane in the VacuCap® 60 and VacuCap® 90 devices provides high flow rates for fast vacuum filtration of 100 ml to 5 litres of aqueous solutions and is available with prefilter to increase throughput of particulate laden solutions.
  • Excellent for cell culture media preparation and available with 0,1 µm pore size to remove mycoplasma
  • Pre-sterilised by gamma irradiation to eliminate contamination by EtO residuals, individually packed
  • Environmentally friendly design with minimal plastic waste