Ansigtsskærme, Clearways
Leverandør: HONEYWELL SAFETY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1002360 1002353 1002354 1002359 1002369 1002366
111-1253EA 122 DKK
111-1253 111-1254 111-3550 111-1359 111-1361 111-1360
Ansigtsskærme, Clearways
Ansigtsskærm
Denne kombination giver både styrke og holdbarhed. Visirene egner sig til forskellige typer af applikationer.
  • Lightweight - comfortable enough to wear all day
  • Integral visor fitting ensures extra protection against chemical seepage
  • Economical replaceable visor available in acetate or PC (PC: High impact; acetate: Chemical resistant)
Vægt: 160 g
Visir: 200 mm
EN 166
Certificeringer: EN 166
Bestillingsoplysninger: Visor, brow guard, and chin guard must be ordered separately. See Accessories tab for full component listings. Holder must also be ordered separately (see 111-1210 or 111-3549 series).