Engangsørepropper, Bilsom 303
Leverandør: HONEYWELL SAFETY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1006186 1006187 1005073 1005074 1007192 1007193 1006204
111-1291EA 406 DKK
111-1291 111-1292 111-3560 111-3564 111-2157 111-2158 DALL1006204
Engangsørepropper, Bilsom 303
Ørepropper Ørepropper til engangsbrug
Formstøbt skumpropper uden PVC, fås i to størrelser for at sikre korrekt og komfortabel pasform.
  • Polyurethane foam, without cord
  • Conical shape for easy insertion
  • Smooth outer surface for better hygiene
  • Transparent dispenser for convenience
SNR: 33 dB
EN 352-2
Certificeringer: EN 352-2.
Emballering: Available bulk packed, as individual pairs or as 400 pairs within the LS-400 dispenser.